Assay Details
Gene Symbol: UTS2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032894.1 | NP_001028066.1 | 360 | 64 | ||
GenBank mRNA | AY065982.1 | - | 360 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
