Assay Details
Gene Symbol: TLR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130429.1 | NP_001123901.1 | 2223 | 97 | ||
XM_015114806.2 | XP_014970292.2 | 2482 | 97 | |||
XM_028851519.1 | XP_028707352.1 | 2320 | 97 | |||
GenBank mRNA | AB445651.1 | - | 2223 | 97 | ||
AY525615.1 | - | 150 | 97 | |||
EU204939.1 | - | 2445 | 97 | |||
JU326826.1 | - | 61 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
