Assay Details
| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130431.2 | NP_001123903.1 | 2715 | 70 | ||
| GenBank mRNA | AB445679.1 | - | 2648 | 70 | ||
| AY525619.1 | - | 44 | 70 | |||
| EU204947.1 | - | 2715 | 70 | |||
| JU476052.1 | - | 2765 | 70 | |||
More Information
Set Membership: |
|

TLR9
Genomic Map