Assay Details
| Gene Symbol: SLCO1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032941.1 | NP_001028113.1 | 507 | 148 | ||
| XM_015151097.2 | XP_015006583.1 | 649 | 148 | |||
| XM_028828782.1 | XP_028684615.1 | 621 | 148 | |||
| GenBank mRNA | AY787037.1 | - | 507 | 148 | ||
More Information
Set Membership: |
|

SLCO1B3
Genomic Map