Assay Details
| Gene Symbol: PRSS23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044737.1 | NP_001038202.1 | 59 | 84 | ||
| XM_028832622.1 | XP_028688455.1 | 451 | 84 | |||
| GenBank mRNA | DQ223039.1 | - | 59 | 84 | ||
| JU471901.1 | - | 152 | 84 | |||
| JU471902.1 | - | 152 | 84 | |||
| JU474130.1 | - | 152 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001044737.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRSS23
Genomic Map