Assay Details
| Gene Symbol: ADRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042774.1 | NP_001036239.1 | 1280 | 78 | ||
| GenBank mRNA | L38905.1 | - | 1280 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRB2| Gene Symbol: ADRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042774.1 | NP_001036239.1 | 1280 | 78 | ||
| GenBank mRNA | L38905.1 | - | 1280 | 78 | ||
Set Membership: |
|