Assay Details
| Gene Symbol: SOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032804.1 | NP_001027976.1 | 167 | 98 | ||
| GenBank mRNA | AB087271.1 | - | 167 | 98 | ||
| JU332147.1 | - | 298 | 98 | |||
| JU472465.1 | - | 283 | 98 | |||
| JV045109.1 | - | 259 | 98 | |||
| JV721782.1 | - | 300 | 98 | |||
More Information
Set Membership: |
|

SOD1
Genomic Map