Assay Details
Gene Symbol: MAMU-DPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048247.1 | NP_001041712.1 | 628 | 100 | ||
GenBank mRNA | AB219099.1 | - | 628 | 100 | ||
EF204948.1 | - | 628 | 100 | |||
HM594293.1 | - | 628 | 100 |
Gene Symbol: LOC114669810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF362455.1 | - | 655 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
