Assay Details
Gene Symbol: MAMU-DPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048247.1 | NP_001041712.1 | 346 | 69 | ||
GenBank mRNA | AB219099.1 | - | 346 | 69 | ||
AB219100.1 | - | 346 | 69 | |||
EF204948.1 | - | 346 | 69 | |||
FJ544416.1 | - | 346 | 69 | |||
HG327295.1 | - | 346 | 69 | |||
HM594293.1 | - | 346 | 69 | |||
JU485417.1 | - | 358 | 69 | |||
JV639240.1 | - | 382 | 69 |
Gene Symbol: LOC114669810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB250756.1 | - | 346 | 69 | ||
HG327296.1 | - | 346 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
