Assay Details
Gene Symbol: MAMU-DPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048247.1 | NP_001041712.1 | 628 | 105 | ||
GenBank mRNA | AB219099.1 | - | 628 | 105 | ||
AB219100.1 | - | 628 | 105 | |||
AB250754.1 | - | 628 | 105 | |||
EF204948.1 | - | 628 | 105 | |||
FJ544416.1 | - | 628 | 105 | |||
HG327295.1 | - | 628 | 105 | |||
HG327297.1 | - | 628 | 105 | |||
HG327299.1 | - | 628 | 105 | |||
HG327302.2 | - | 628 | 105 | |||
HM594293.1 | - | 628 | 105 | |||
JV639240.1 | - | 664 | 105 |
Gene Symbol: LOC114669810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB250756.1 | - | 628 | 105 | ||
EF204950.1 | - | 628 | 105 | |||
EF362455.1 | - | 655 | 105 | |||
HG327296.1 | - | 628 | 105 | |||
JU334259.1 | - | 654 | 105 | |||
LN875388.1 | - | 628 | 105 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
