Assay Details
Gene Symbol: MAMU-DPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048247.1 | NP_001041712.1 | 632 | 62 | ||
XM_015136066.2 | XP_014991552.2 | 1422 | 62 | |||
GenBank mRNA | AB219099.1 | - | 632 | 62 | ||
AB219100.1 | - | 632 | 62 | |||
AB250754.1 | - | 632 | 62 | |||
EF204948.1 | - | 632 | 62 | |||
EF362456.1 | - | 632 | 62 | |||
EU305663.1 | - | 632 | 62 | |||
EU934786.1 | - | 632 | 62 | |||
FJ009192.1 | - | 632 | 62 | |||
FJ544416.1 | - | 632 | 62 | |||
HG007852.1 | - | 632 | 62 | |||
HG327295.1 | - | 632 | 62 | |||
HG327297.1 | - | 632 | 62 | |||
HG327299.1 | - | 632 | 62 | |||
HG327302.2 | - | 632 | 62 | |||
HG514471.1 | - | 632 | 62 | |||
HM594293.1 | - | 632 | 62 | |||
JU485417.1 | - | 644 | 62 | |||
JV639240.1 | - | 668 | 62 |
Gene Symbol: LOC114669810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842593.1 | XP_028698426.1 | 840 | 62 | ||
GenBank mRNA | AB250756.1 | - | 632 | 62 | ||
EF204947.1 | - | 632 | 62 | |||
EF204950.1 | - | 632 | 62 | |||
EF362455.1 | - | 659 | 62 | |||
HG327296.1 | - | 632 | 62 | |||
JU334259.1 | - | 658 | 62 | |||
LN875388.1 | - | 632 | 62 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
