Assay Details
Gene Symbol: PPARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032860.1 | NP_001028032.1 | 835 | 140 | ||
XM_015130199.2 | XP_014985685.1 | 1127 | 140 | |||
XM_015130200.2 | XP_014985686.1 | 985 | 140 | |||
XM_015130202.2 | XP_014985688.1 | 762 | 140 | |||
XM_028843038.1 | XP_028698871.1 | 1210 | 140 | |||
XM_028843040.1 | XP_028698873.1 | 1276 | 140 | |||
XM_028843041.1 | XP_028698874.1 | 1276 | 140 | |||
XM_028843042.1 | XP_028698875.1 | 1222 | 140 | |||
XM_028843043.1 | XP_028698876.1 | 1153 | 140 | |||
XM_028843044.1 | XP_028698877.1 | 1203 | 140 | |||
GenBank mRNA | AF033103.1 | - | 835 | 140 | ||
AF033342.1 | - | 870 | 140 | |||
AF033343.1 | - | 842 | 140 | |||
JV654393.1 | - | 158 | 140 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
