Assay Details
| Gene Symbol: ABO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116452.2 | XP_014971938.2 | 607 | 65 | ||
| XM_015116453.2 | XP_014971939.2 | 559 | 65 | |||
| XM_015116454.2 | XP_014971940.2 | 452 | 65 | |||
| XM_015116455.2 | XP_014971941.1 | 404 | 65 | |||
| XM_028834648.1 | XP_028690481.1 | 610 | 65 | |||
| XM_028834649.1 | XP_028690482.1 | 449 | 65 | |||
| XM_028834650.1 | XP_028690483.1 | 401 | 65 | |||
| XM_028834651.1 | XP_028690484.1 | 1017 | 65 | |||
| XM_028834652.1 | XP_028690485.1 | 1014 | 65 | |||
| GenBank mRNA | AF071830.1 | - | 212 | 65 | ||
| JV503238.1 | - | 373 | 65 | |||
| JV578799.1 | - | 247 | 65 | |||
More Information
Set Membership: |
|

ABO
Genomic Map