Assay Details
| Gene Symbol: ESR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265821.2 | NP_001252750.1 | 1506 | 117 | ||
| XM_015143836.2 | XP_014999322.2 | 2114 | 117 | |||
| GenBank mRNA | AF119229.1 | - | 377 | 117 | ||
| JV636022.1 | - | 1509 | 117 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ESR2
Genomic Map