Assay Details
| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032814.1 | NP_001027986.1 | 200 | 70 | ||
| GenBank mRNA | AF162280.1 | - | 200 | 70 | ||
| JU322553.1 | - | 269 | 70 | |||
| JV529872.1 | - | 267 | 70 | |||
More Information
Set Membership: |
|

NPY
Genomic Map