Assay Details
| Gene Symbol: VAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032820.2 | NP_001027992.1 | 436 | 62 | ||
| GenBank mRNA | AF240769.1 | - | 430 | 62 | ||
| JV044681.1 | - | 423 | 62 | |||
| JV565238.1 | - | 435 | 62 | |||
| JV635626.1 | - | 438 | 62 | |||
| JV669158.1 | - | 436 | 62 | |||
More Information
Set Membership: |
|

VAMP2
Genomic Map