Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032834.2 | NP_001028006.1 | 223 | 102 | ||
| XM_015115661.2 | XP_014971147.1 | 1169 | 102 | |||
| XM_015115662.2 | XP_014971148.1 | 1258 | 102 | |||
| XM_015115663.2 | XP_014971149.1 | 1157 | 102 | |||
| GenBank mRNA | AF303732.1 | - | 223 | 102 | ||
| JV044311.1 | - | 305 | 102 | |||
| JV639328.1 | - | 326 | 102 | |||
More Information
Set Membership: |
|

IL18
Genomic Map