Assay Details
| Gene Symbol: GNRHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032842.1 | NP_001028014.1 | 528 | 93 | ||
| GenBank mRNA | AF353987.1 | - | 528 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNRHR2| Gene Symbol: GNRHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032842.1 | NP_001028014.1 | 528 | 93 | ||
| GenBank mRNA | AF353987.1 | - | 528 | 93 | ||
Set Membership: |
|