Assay Details
| Gene Symbol: UGT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032869.1 | NP_001028041.1 | 863 | 79 | ||
| GenBank mRNA | AF360121.1 | - | 863 | 79 | ||
| AY029169.1 | - | 863 | 79 | |||
More Information
Set Membership: |
|

UGT1A1
Genomic Map