Assay Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033089.1 | NP_001028261.1 | 48 | 86 | ||
| GenBank mRNA | AF369755.1 | - | 48 | 86 | ||
| AY040319.1 | - | 59 | 86 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD209
Genomic Map