Assay Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032870.1 | NP_001028042.1 | 120 | |||
| NM_001033089.1 | NP_001028261.1 | 684 | 120 | |||
| GenBank mRNA | AF369755.1 | - | 684 | 120 | ||
| AF391086.1 | - | 120 | ||||
| AY040319.1 | - | 695 | 120 | |||
| JU485348.1 | - | 120 | ||||
More Information
Set Membership: |
|

CD209
Genomic Map