Assay Details
| Gene Symbol: CCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034200.1 | NP_001029372.1 | 186 | 54 | ||
| GenBank mRNA | AF449266.1 | - | 186 | 54 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL3| Gene Symbol: CCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034200.1 | NP_001029372.1 | 186 | 54 | ||
| GenBank mRNA | AF449266.1 | - | 186 | 54 | ||
Set Membership: |
|