Assay Details
| Gene Symbol: CCL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032853.1 | NP_001028025.1 | 70 | 89 | ||
| GenBank mRNA | AF449272.1 | - | 70 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL18| Gene Symbol: CCL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032853.1 | NP_001028025.1 | 70 | 89 | ||
| GenBank mRNA | AF449272.1 | - | 70 | 89 | ||
Set Membership: |
|