Assay Details
| Gene Symbol: CCL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032854.1 | NP_001028026.1 | 188 | 67 | ||
| XM_015110957.2 | XP_014966443.1 | 363 | 67 | |||
| GenBank mRNA | AF449274.1 | - | 188 | 67 | ||
More Information
Set Membership: |
|

CCL20
Genomic Map