Assay Details
| Gene Symbol: CCL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032946.1 | NP_001028118.1 | 132 | 89 | ||
| XM_028835457.1 | XP_028691290.1 | 347 | 89 | |||
| GenBank mRNA | AF449276.1 | - | 132 | 89 | ||
More Information
Set Membership: |
|

CCL23
Genomic Map