Assay Details
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032943.1 | NP_001028115.1 | 105 | 102 | ||
XM_015150991.2 | XP_015006477.2 | 292 | 102 | |||
XM_015150992.2 | XP_015006478.2 | 292 | 102 | |||
XM_015150993.2 | XP_015006479.2 | 292 | 102 | |||
GenBank mRNA | AF508729.1 | - | 105 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
