Assay Details
Gene Symbol: MAMU-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ601348.1 | - | 366 | 154 | ||
AJ601350.1 | - | 366 | 154 | |||
EU934774.1 | - | 366 | 154 | |||
GQ471892.1 | - | 366 | 154 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
