Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042658.1 | NP_001036123.1 | 668 | 139 | ||
| GenBank mRNA | AY094979.1 | - | 668 | 139 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD1E| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042658.1 | NP_001036123.1 | 668 | 139 | ||
| GenBank mRNA | AY094979.1 | - | 668 | 139 | ||
Set Membership: |
|