Assay Details
| Gene Symbol: NR1I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032896.1 | NP_001028068.1 | 719 | 95 | ||
| GenBank mRNA | AY116212.1 | - | 719 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR1I3| Gene Symbol: NR1I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032896.1 | NP_001028068.1 | 719 | 95 | ||
| GenBank mRNA | AY116212.1 | - | 719 | 95 | ||
Set Membership: |
|