Assay Details
| Gene Symbol: HRH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032898.1 | NP_001028070.1 | 376 | 56 | ||
| XM_015148635.2 | XP_015004121.1 | 551 | 56 | |||
| GenBank mRNA | AY231164.1 | - | 376 | 56 | ||
| JV601301.1 | - | 552 | 56 | |||
More Information
Set Membership: |
|

HRH3
Genomic Map