Assay Details
| Gene Symbol: APOBEC3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198693.2 | NP_001185622.1 | 861 | 109 | ||
| GenBank mRNA | AY331716.1 | - | 560 | 109 | ||
| DQ913647.1 | - | 553 | 109 | |||
| DQ913648.1 | - | 547 | 109 | |||
| DQ913649.1 | - | 476 | 109 | |||
| DQ913650.1 | - | 551 | 109 | |||
| DQ913651.1 | - | 406 | 109 | |||
| DQ913652.1 | - | 527 | 109 | |||
| DQ913653.1 | - | 547 | 109 | |||
| DQ913654.1 | - | 406 | 109 | |||
| DQ913655.1 | - | 488 | 109 | |||
| DQ913656.1 | - | 547 | 109 | |||
| DQ913657.1 | - | 547 | 109 | |||
| DQ913658.1 | - | 547 | 109 | |||
| DQ913659.1 | - | 547 | 109 | |||
| DQ913660.1 | - | 544 | 109 | |||
| DQ913661.1 | - | 547 | 109 | |||
| DQ913662.1 | - | 547 | 109 | |||
| DQ913663.1 | - | 204 | 109 | |||
| DQ913664.1 | - | 547 | 109 | |||
| DQ913665.1 | - | 408 | 109 | |||
| DQ913666.1 | - | 406 | 109 | |||
| DQ913667.1 | - | 526 | 109 | |||
| DQ913668.1 | - | 552 | 109 | |||
| DQ913669.1 | - | 553 | 109 | |||
| DQ913670.1 | - | 406 | 109 | |||
| DQ913671.1 | - | 547 | 109 | |||
| DQ913672.1 | - | 526 | 109 | |||
| DQ913673.1 | - | 406 | 109 | |||
| DQ913674.1 | - | 410 | 109 | |||
| DQ913675.1 | - | 406 | 109 | |||
| DQ913676.1 | - | 547 | 109 | |||
| DQ913677.1 | - | 523 | 109 | |||
| DQ913678.1 | - | 547 | 109 | |||
| DQ913679.1 | - | 544 | 109 | |||
| DQ913680.1 | - | 524 | 109 | |||
| DQ913681.1 | - | 408 | 109 | |||
| DQ913682.1 | - | 405 | 109 | |||
| DQ913683.1 | - | 552 | 109 | |||
| DQ913684.1 | - | 547 | 109 | |||
| DQ913685.1 | - | 410 | 109 | |||
| DQ913686.1 | - | 406 | 109 | |||
| DQ913687.1 | - | 524 | 109 | |||
| DQ913688.1 | - | 408 | 109 | |||
| DQ913689.1 | - | 406 | 109 | |||
| DQ913690.1 | - | 526 | 109 | |||
| JU491021.1 | - | 571 | 109 | |||
| JX915798.1 | - | 584 | 109 | |||
| KC176183.1 | - | 581 | 109 | |||
| KC176184.1 | - | 581 | 109 | |||
| KF020481.1 | - | 584 | 109 | |||
| KF020482.1 | - | 584 | 109 | |||
| KF020483.1 | - | 584 | 109 | |||
| KF020484.1 | - | 584 | 109 | |||
| KF020485.1 | - | 584 | 109 | |||
| KF020486.1 | - | 581 | 109 | |||
| KU058146.1 | - | 788 | 109 | |||
| KU058147.2 | - | 617 | 109 | |||
More Information
Set Membership: |
|

APOBEC3G
Genomic Map