Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032930.1 | NP_001028102.1 | 1329 | 63 | ||
| GenBank mRNA | AY340665.1 | - | 1329 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCC13| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032930.1 | NP_001028102.1 | 1329 | 63 | ||
| GenBank mRNA | AY340665.1 | - | 1329 | 63 | ||
Set Membership: |
|