Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195417.1 | NP_001182346.1 | 525 | 68 | ||
GenBank mRNA | AY369789.1 | - | 171 | 68 | ||
CO726823.1 | - | 569 | 68 | |||
JU336288.1 | - | 656 | 68 |

More Information
Set Membership: |
![]() ![]() |
