Assay Details
| Gene Symbol: MACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038650.1 | NP_001033739.1 | 1904 | 87 | ||
| GenBank mRNA | AY845017.1 | - | 1904 | 87 | ||
| AY845038.1 | - | 733 | 87 | |||
| JU335272.1 | - | 1825 | 87 | |||
| JU472111.1 | - | 1854 | 87 | |||
| JV043299.1 | - | 1831 | 87 | |||
| JV586220.1 | - | 1880 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MACO1
Genomic Map