Assay Details
| Gene Symbol: NPSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032942.1 | NP_001028114.1 | 151 | 84 | ||
| GenBank mRNA | AY950595.1 | - | 151 | 84 | ||
| AY956445.1 | - | 151 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NPSR1
Genomic Map