Assay Details
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266703.1 | NP_001253632.1 | 125 | 74 | ||
XM_028853380.1 | XP_028709213.1 | 435 | 74 | |||
XM_028853383.1 | XP_028709216.1 | 440 | 74 | |||
XM_028853384.1 | XP_028709217.1 | 440 | 74 | |||
GenBank mRNA | DQ148090.1 | - | 87 | 74 | ||
JU333277.1 | - | 129 | 74 | |||
JV045160.1 | - | 125 | 74 | |||
JV573672.1 | - | 150 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
