Assay Details
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195502.1 | NP_001182431.1 | 54 | 69 | ||
GenBank mRNA | CD767990.1 | - | 54 | 69 | ||
DQ148153.1 | - | 42 | 69 | |||
JV722244.1 | - | 67 | 69 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
