Assay Details
| Gene Symbol: LILRB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040676.1 | NP_001035766.1 | 1031 | 138 | ||
| GenBank mRNA | DQ155433.1 | - | 1031 | 138 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LILRB4| Gene Symbol: LILRB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040676.1 | NP_001035766.1 | 1031 | 138 | ||
| GenBank mRNA | DQ155433.1 | - | 1031 | 138 | ||
Set Membership: |
|