Assay Details
| Gene Symbol: LOC698874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849774.1 | XP_028705607.1 | 708 | 142 | ||
| XM_028849779.1 | XP_028705612.1 | 827 | 142 | |||
More Information
Set Membership: |
|

LOC698874
Genomic Map