Assay Details
Gene Symbol: PTP4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266125.1 | NP_001253054.1 | 923 | 157 | ||
GenBank mRNA | DQ147968.1 | - | 280 | 157 | ||
JU320043.1 | - | 826 | 157 | |||
JU471059.1 | - | 923 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
