Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042666.1 | NP_001036131.1 | 471 | 80 | ||
| XM_015149359.2 | XP_015004845.1 | 638 | 80 | |||
| XM_015149360.2 | XP_015004846.1 | 425 | 80 | |||
| XM_028827424.1 | XP_028683257.1 | 625 | 80 | |||
| GenBank mRNA | DQ842019.1 | - | 471 | 80 | ||
| JU335628.1 | - | 593 | 80 | |||
| JU335629.1 | - | 593 | 80 | |||
| JU335630.1 | - | 593 | 80 | |||
| JU476541.1 | - | 586 | 80 | |||
| JV047759.1 | - | 567 | 80 | |||
| JV656977.1 | - | 603 | 80 | |||
More Information
Set Membership: |
|

MAVS
Genomic Map