Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032813.1 | NP_001027985.1 | 846 | 127 | ||
| XM_015135838.2 | XP_014991324.2 | 992 | 127 | |||
| GenBank mRNA | AF147880.1 | - | 846 | 127 | ||
| JU482220.1 | - | 901 | 127 | |||
| JV046412.1 | - | 814 | 127 | |||
| JV046414.1 | - | 814 | 127 | |||
More Information
Set Membership: |
|

TUBB
Genomic Map