Assay Details
| Gene Symbol: GH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042738.1 | NP_001036203.1 | 536 | 145 | ||
| XM_015120065.2 | XP_014975551.2 | 690 | 145 | |||
| XM_015120066.2 | XP_014975552.1 | 574 | 145 | |||
| GenBank mRNA | L16556.1 | - | 536 | 145 | ||
More Information
Set Membership: |
|

GH1
Genomic Map