Assay Details
Gene Symbol: ANAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260774.1 | NP_001247703.1 | 759 | 113 | ||
XM_028834298.1 | XP_028690131.1 | 826 | 113 | |||
GenBank mRNA | JU334200.1 | - | 772 | 113 | ||
JU475326.1 | - | 761 | 113 | |||
JV046876.1 | - | 759 | 113 | |||
JV681874.1 | - | 777 | 113 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
