Assay Details
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195379.1 | NP_001182308.1 | 249 | 64 | ||
GenBank mRNA | CB308101.1 | - | 242 | 64 | ||
DV769814.2 | - | 253 | 64 | |||
JU323400.1 | - | 237 | 64 | |||
JU471700.1 | - | 229 | 64 | |||
JV047099.1 | - | 261 | 64 | |||
JV651787.1 | - | 235 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
