Assay Details
Gene Symbol: LOC715257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110742.2 | XP_014966228.2 | 550 | 88 | ||
XM_015110743.2 | XP_014966229.2 | 590 | 88 | |||
XM_015110744.2 | XP_014966230.2 | 413 | 88 | |||
XM_015110745.2 | XP_014966231.2 | 292 | 88 | |||
GenBank mRNA | JU476529.1 | - | 323 | 88 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
