Assay Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265636.2 | NP_001252565.1 | 356 | 72 | ||
| XM_015133521.2 | XP_014989007.1 | 864 | 72 | |||
| XM_015133522.2 | XP_014989008.1 | 445 | 72 | |||
| XM_015133523.2 | XP_014989009.1 | 796 | 72 | |||
| GenBank mRNA | JU335415.1 | - | 360 | 72 | ||
| JU476360.1 | - | 362 | 72 | |||
| JV047615.1 | - | 316 | 72 | |||
| JV565148.1 | - | 358 | 72 | |||
| JV621902.1 | - | 362 | 72 | |||
More Information
Set Membership: |
|

COA1
Genomic Map