Assay Details
| Gene Symbol: ATP6V0E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261126.1 | NP_001248055.1 | 173 | 69 | ||
| GenBank mRNA | JU321368.1 | - | 163 | 69 | ||
| JU473429.1 | - | 173 | 69 | |||
More Information
Set Membership: |
|

ATP6V0E1
Genomic Map