Assay Details
| Gene Symbol: GTF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095556.4 | XP_001095556.2 | 1127 | 90 | ||
| GenBank mRNA | JU342469.1 | - | 1061 | 90 | ||
| JV681828.1 | - | 1060 | 90 | |||
More Information
Set Membership: |
|

GTF3A
Genomic Map