Assay Details
Gene Symbol: LOC694289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083221.4 | XP_001083221.2 | 2966 | 57 | ||
XM_015148704.2 | XP_015004190.2 | 2969 | 57 | |||
XM_015148705.2 | XP_015004191.2 | 2924 | 57 | |||
XM_015148706.2 | XP_015004192.2 | 2921 | 57 | |||
XM_015148707.2 | XP_015004193.1 | 540 | 57 | |||
XM_015148708.2 | XP_015004194.1 | 407 | 57 | |||
XM_028828531.1 | XP_028684364.1 | 822 | 57 | |||
XM_028828532.1 | XP_028684365.1 | 858 | 57 | |||
XM_028828533.1 | XP_028684366.1 | 777 | 57 | |||
XM_028828534.1 | XP_028684367.1 | 813 | 57 | |||
XM_028828535.1 | XP_028684368.1 | 361 | 57 | |||
XM_028828536.1 | XP_028684369.1 | 1456 | 57 | |||
XM_028828537.1 | XP_028684370.1 | 338 | 57 | |||
XM_028828538.1 | XP_028684371.1 | 460 | 57 | |||
XM_028828539.1 | XP_028684372.1 | 518 | 57 | |||
XM_028828540.1 | XP_028684373.1 | 350 | 57 | |||
GenBank mRNA | JU477879.1 | - | 935 | 57 | ||
JV043979.1 | - | 480 | 57 | |||
JV043980.1 | - | 480 | 57 | |||
JV600239.1 | - | 411 | 57 | |||
JV600240.1 | - | 414 | 57 | |||
JV712359.1 | - | 690 | 57 |
Gene Symbol: LOC100424899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148709.2 | XP_015004195.1 | 1493 | 57 | ||
XM_015148710.2 | XP_015004196.1 | 1448 | 57 |
Gene Symbol: TADA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU477880.1 | - | 935 | 57 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
